Ponceau S 0,2% v/v acetic acid_5

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt, 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, 3-Hydroxy-4-((2-sulfo-4-((4-sulfophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulfonic acid, tetrasodium salt, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt

J63139-AP J63139-K2
CAAAJ63139-APEA 175.14 CAD
CAAAJ63139-AP CAAAJ63139-K2
Ponceau S 0,2% v/v acetic acid_5
Ponceau S

Liquid


CAS No.: 6226-79-5 Molecular Formula: C22H12N4Na4O13S4 Formula Weight: 760.56 Physical Form: Liquid Appearance: Red MDL No.: MFCD00003892

Formula: C₂₂H₁₂N₄Na₄O₁₃S₄
MW: 760.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003892
CAS Number: 6226-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR