(+)-Dehydroabietylamine hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J63123-MA J63123-MF
CAAAJ63123-MAEA 336.21 CAD
CAAAJ63123-MA CAAAJ63123-MF
(+)-Dehydroabietylamine hydrochloride
(+)-Dehydroabietylamine hydrochloride

Resolving agent for carboxylic acids


White solid


Applications: Resolving agent for carboxylic acids

Formula: C₂₀H₃₁N·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00213430
CAS Number: 16496-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR