Triflumuron 98+%

Supplier: Thermo Scientific Chemicals

J63112-09 J63112-14 J63112-22
CAAAJ63112-09EA 180.53 CAD
CAAAJ63112-09 CAAAJ63112-14 CAAAJ63112-22
Triflumuron 98+%
Triflumuron

Triflumuron acts as a chitin synthesis inhibitor and prevents uridine incorporation into RNA


Solid


Triflumuron acts as a chitin synthesis inhibitor and prevents uridine incorporation into RNA

Formula: C₁₅H₁₀ClF₃N₂O₃
MW: 358.7 g/mol
Melting Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00072496
CAS Number: 64628-44-0
EINECS: 264-980-3
Merck Index: 14,9678

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR