Naproxen sodium 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
J63103-06
J63103-14
J63103-22
CAAAJ63103-06EA
164.23
CAD
CAAAJ63103-06
CAAAJ63103-14
CAAAJ63103-22
Naproxen sodium 98%
Naproxen sodium
A cyclooxygenase inhibitor
Powder
Applications: A cyclooxygenase inhibitor
Formula:
C₁₄H₁₃NaO₃ Melting Pt: 259…262 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058507 CAS Number: 26159-34-2 EINECS: 247-486-2 Merck Index: 14,6417 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...