Ornidazole 99%

Supplier: Thermo Scientific Chemicals
Warning

J63100-06 J63100-14
CAAAJ63100-06EA 155.55 CAD
CAAAJ63100-06 CAAAJ63100-14
Ornidazole 99%
Ornidazole

Interferes with electron transport in metabolic pathways


Powder


Applications: Interferes with electron transport in metabolic pathways

Formula: C₇H₁₀ClN₃O₃
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00057960
CAS Number: 16773-42-5
EINECS: 240-826-0
Merck Index: 14,6872

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR