20-Hydroxyecdysone

Supplier: Thermo Scientific Chemicals

Synonyms: β-Ecdysone

J63091-MA J63091-MF
CAAAJ63091-MAEA 535.13 CAD
CAAAJ63091-MA CAAAJ63091-MF
20-Hydroxyecdysone
20-Hydroxyecdysone

An insect steroid hormone that induces apoptosis


Powder


Applications: An insect steroid hormone that induces apoptosis

Formula: C₂₇H₄₄O₇
Melting Pt: 242…244 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036740
CAS Number: 5289-74-7
Merck Index: 14,3491

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR