Polymyxin B sulphate

Supplier: Thermo Scientific Chemicals
Warning

J63074-03 J63074-06
CAAAJ63074-03EA 336.59 CAD
CAAAJ63074-03 CAAAJ63074-06
Polymyxin B sulphate
Polymyxin B sulphate

Antibiotic for Gram-negative bacteria


White to pale yellow powder


Soluble in water at 50mg/ml

Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00131991
CAS Number: 1405-20-5
EINECS: 215-774-7
Merck Index: 14,7573

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR