Nabumetone

Supplier: Thermo Scientific Chemicals

J63072-06 J63072-14
CAAAJ63072-06EA 267.81 CAD
CAAAJ63072-06 CAAAJ63072-14
Nabumetone
Nabumetone

Anti-inflammatory, anti-bacterial. A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase


Crystalline powder


Applications: Anti-inflammatory, anti-bacterial. A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase

Formula: C₁₅H₁₆O₂
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00079518
CAS Number: 42924-53-8
Merck Index: 14,6343

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR