L-Cystine disodium salt monohydrate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (H-Cys-OH)2.2Na.H2O

J63071-18 J63071-30
CAAAJ63071-18EA 156.53 CAD
CAAAJ63071-18 CAAAJ63071-30
L-Cystine disodium salt monohydrate ≥98%
L-Cystine disodium salt monohydrate

Powder


Purity: 98+% CAS No.: 199329-53-8 Molecular Formula: C6H10N2Na2O4S2.H2O Formula Weight: 302.27 (284.26anhy) Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00243291

Formula: C₆H₁₀N₂Na₂O₄S₂·H₂O
MW: 302.27 g/mol (284.26 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00243291
CAS Number: 199329-53-8
EINECS: 265-025-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR