Trazodone hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J63070-03 J63070-06
CAAAJ63070-03EA 147.16 CAD
CAAAJ63070-03 CAAAJ63070-06
Trazodone hydrochloride
Trazodone hydrochloride

A serotonin uptake inhibitor


Powder


Applications: A serotonin uptake inhibitor

Formula: C₁₉H₂₃Cl₂N₅O
Melting Pt: 222…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00079603
CAS Number: 25332-39-2
EINECS: 246-855-5
Merck Index: 14,9579

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR