Spermine tetrahydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine) tetrahydrochloride

J63060-03 J63060-06 J63060-14
CAAAJ63060-03EA 72.13 CAD
CAAAJ63060-03 CAAAJ63060-06 CAAAJ63060-14
Spermine tetrahydrochloride 99%
Spermine tetrahydrochloride

Binds to the polyamine modulatory site of NMDA.


Powder


Binds to the polyamine modulatory site of NMDA

Formula: C₁₀H₃₀Cl₄N₄
Storage Temperature: Ambient
MDL Number: MFCD00012914
CAS Number: 306-67-2
EINECS: 206-189-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR