Capsazepine ≥99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-(4-Chlorophenethyl)-7,8-dihydroxy-4,5-dihydro-1H-benzo[c]azepine-2(3H)-carbothioamide

J63055-MA J63055-MF
CAAAJ63055-MAEA 467.87 CAD
CAAAJ63055-MA CAAAJ63055-MF
Capsazepine ≥99%
Capsazepine

A competitive antagonist for capsaicin and resiniferatoxin


Powder


Applications: A competitive antagonist for capsaicin and resiniferatoxin

Formula: C₁₉H₂₁ClN₂O₂S
MW: 376.90 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00153778
CAS Number: 138977-28-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR