Neutral red
Supplier: Thermo Scientific Chemicals
J63054-03
J63054-06
J63054-14
CAAAJ63054-03EA
70.48
CAD
CAAAJ63054-03
CAAAJ63054-06
CAAAJ63054-14
Neutral red
Neutral red
Nuclear counterstain useful in pH range 6.8 - 8.0.
Powder
Nuclear counterstain useful in pH range 6.8 - 8.0
Formula:
C₁₅H₁₇ClN₄ Melting Pt: 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012651 CAS Number: 553-24-2 EINECS: 209-035-8 Merck Index: 14,6488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...