Trimethoprim

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine

J63053-03 J63053-06 J63053-14
CAAAJ63053-03EA 81.09 CAD
CAAAJ63053-03 CAAAJ63053-06 CAAAJ63053-14
Trimethoprim
Trimethoprim

An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme


Powder


Applications: An antibacterial and inhibitor of formylation. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme

Formula: C₁₄H₁₈N₄O₃
Melting Pt: 199…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00036761
CAS Number: 738-70-5
EINECS: 212-006-2
Merck Index: 14,9709

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR