L(+)-Isoleucine cell culture reagent
Supplier: Thermo Scientific Chemicals
J63045-14
J63045-22
CAAAJ63045-14EA
153.01
CAD
CAAAJ63045-14
CAAAJ63045-22
L(+)-Isoleucine cell culture reagent
L(+)-Isoleucine
Powder
CAS No.: 73-32-5
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Physical Form: Powder
Appearance: White to off-white
Melting Point: ca 287° dec.
Beilstein Reference: 1721792
Merck Reference: 14,5179
Optical Rotation: +40° (c=5 in 6N HCl)
MDL No.: MFCD00064222
Formula:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Melting Pt: ∼ 287 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064222 CAS Number: 73-32-5 EINECS: 200-798-2 Merck Index: 14,5179 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...