Penicillin G sodium salt

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

J63032-06 J63032-14 J63032-22
CAAAJ63032-06EA 49 CAD
CAAAJ63032-06 CAAAJ63032-14 CAAAJ63032-22
Penicillin G sodium salt
Penicillin G sodium salt

Inhibits bacterial cell wall synthesis


Powder


Inhibits bacterial cell wall synthesis

Formula: C₁₆H₁₇N₂NaO₄S
Melting Pt: 255 °C
Density: 1.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00069666
CAS Number: 69-57-8
EINECS: 200-710-2
Merck Index: 14,7094

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR