Sodium Bicarbonate, 1 M Buffer Solution, pH 10.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J63025-AK J63025-AP
CAAAJ63025-AKEA 135.09 CAD
CAAAJ63025-AK CAAAJ63025-AP
Sodium Bicarbonate, 1 M Buffer Solution, pH 10.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 144-55-8
Physical Form: Liquid
MDL No.: MFCD00003528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR