Vitamin A palmitate, 1.7 MIU/g
Supplier: Thermo Scientific Chemicals
|
Danger
|
J63022-14
J63022-22
CAAAJ63022-14EA
152.72
CAD
CAAAJ63022-14
CAAAJ63022-22
Vitamin A palmitate, 1.7 MIU/g
Vitamin A palmitate
CAS No.: 79-81-2
Molecular Formula: C36H60O2
Formula Weight: 524.87
Storage Temperature: 2°C to 8°C
Physical Form: Low melting solid
Appearance: Yellow to orange
Melting Point: 28-29°
Beilstein Reference: 1917366
Merck Reference: 14,10013
MDL No.: MFCD00019414
Formula:
C₃₆H₆₀O₂ Melting Pt: 28…29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019414 CAS Number: 79-81-2 EINECS: 201-228-5 Merck Index: 14,10013 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...