Phosphoenol Pyruvic acidTrisodium Salt Hydrate 98%
Supplier: Thermo Scientific Chemicals
J63013-03
J63013-06
CAAAJ63013-03EA
367.27
CAD
CAAAJ63013-03
CAAAJ63013-06
Phosphoenol Pyruvic acidTrisodium Salt Hydrate 98%
Phosphoenol pyruvic acid trisodium salt hydrate
When coupled with pyruvate kinase it completes an ATP-regenerating system
Powder
Applications: When coupled with pyruvate kinase it completes an ATP-regenerating system
Formula:
C₃H₂Na₃O₆P MW: 233.986 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150737 CAS Number: 5541-93-5 EINECS: 226-906-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...