DL-Adrenaline
Supplier: Thermo Scientific Chemicals
|
Danger
|
J63005-03
J63005-14
CAAAJ63005-03EA
55.3
CAD
CAAAJ63005-03
CAAAJ63005-14
DL-Adrenaline
DL-Adrenaline
A hormone and neurotransmitter
CAS No.: 329-65-7
Molecular Formula: C9H13NO3
Formula Weight: 183.20
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Off-white
Melting Point: 197° dec.
UN No.: 2811
MDL No.: MFCD00063027
Applications: A hormone and neurotransmitter
Formula:
C₉H₁₃NO₃ Melting Pt: 197 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063027 CAS Number: 329-65-7 EINECS: 206-347-6 UN: 2811 ADR: "6.1",III Merck Index: 14,3619 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...