1-Naphthyl phosphate monosodium salt monohydrate 97+%

Supplier: Thermo Scientific Chemicals
Warning

J63004-14 J63004-06
CAAAJ63004-14EA 1415.74 CAD
CAAAJ63004-14 CAAAJ63004-06
1-Naphthyl phosphate monosodium salt monohydrate 97+%
1-Naphthyl phosphate monosodium salt monohydrate

Substrate for determination of phosphatases.


Powder


Substrate for determination of phosphatases

Formula: C₁₀H₈NaO₄P·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150615
CAS Number: 81012-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR