Tobramycin sulfate

Supplier: Thermo Scientific Chemicals
Warning

J62995-03 J62995-MC
CAAAJ62995-03EA 917.77 CAD
CAAAJ62995-03 CAAAJ62995-MC
Tobramycin sulfate
Tobramycin sulfate

A water-soluble aminoglycoside antibiotic


Powder


Applications: A water-soluble aminoglycoside antibiotic

Formula: (C₁₈H₃₇N₅O₉)₂·5H₂SO₄
MW: 1425.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00133864
CAS Number: 79645-27-5
Merck Index: 14,9490

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR