RBC lysis buffer for human, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62990-AK
J62990-AP
CAAAJ62990-AKEA
119.4
CAD
CAAAJ62990-AK
CAAAJ62990-AP
RBC lysis buffer for human, Thermo Scientific
Buffers
Extraction Buffers
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Applications: For lysis of human red blood cells.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...