EDTA (ethylenediamine tetraacetic acid) cell culture reagent
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
J62948-36
J62948-A1
CAAAJ62948-36EA
179.11
CAD
CAAAJ62948-36
CAAAJ62948-A1
EDTA (ethylenediamine tetraacetic acid) cell culture reagent
EDTA (Ethylenediamine tetraacetic acid)
Powder
CAS No.: 60-00-4
Molecular Formula: C10H16N2O8
Formula Weight: 292.24
Physical Form: Powder
Appearance: White
Melting Point: ca 245° dec.
Beilstein Reference: 1716295
Merck Reference: 14,3517
MDL No.: MFCD00003541
Formula:
C₁₀H₁₆N₂O₈ Boiling Pt: 614 °C (1013 hPa) Melting Pt: ∼ 245 °C (dec.) Density: 0.86 g/cm³ (20 °C) Flash Pt: ≥ 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 REACH: 01-2119486399-18 Merck Index: 14,3517 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...