TRIS-buffered saline (TBS, 10X) pH 7.4, for Western blot, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62938-K2 J62938-K3 J62938-K7
CAAAJ62938-K2EA 105.23 CAD
CAAAJ62938-K2 CAAAJ62938-K3 CAAAJ62938-K7
TRIS-buffered saline (TBS, 10X) pH 7.4, for Western blot, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Liquid


CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00004679
Applications: For Western blot washing.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR