Folic acid cell culture reagent

Supplier: Thermo Scientific Chemicals

J62937-06 J62937-14 J62937-22
CAAAJ62937-06EA 63.77 CAD
CAAAJ62937-06 CAAAJ62937-14 CAAAJ62937-22
Folic acid cell culture reagent
Folic acid

Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells


Powder


Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells

Formula: C₁₉H₁₉N₇O₆
Storage Temperature: Ambient
MDL Number: MFCD00079305
CAS Number: 59-30-3
EINECS: 200-419-0
Merck Index: 14,4221

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR