Neohesperidin dihydrochalcone hydrate ≥98%

Supplier: Thermo Scientific Chemicals

J62922-06 J62922-14 J62922-03
CAAAJ62922-06EA 208.31 CAD
CAAAJ62922-06 CAAAJ62922-14 CAAAJ62922-03
Neohesperidin dihydrochalcone hydrate ≥98%
Neohesperidin dihydrochalcone hydrate

Artificial sweetener derived from citrus


Crystalline powder


Applications: Artificial sweetener derived from citrus

Formula: C₂₈H₃₆O₁₅.xH₂O
Storage Temperature: Ambient
MDL Number: MFCD03840557
CAS Number: 20702-77-6
EINECS: 243-978-6
Merck Index: 14,6452

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR