Glycylglycine, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62920-AE
J62920-AK
CAAAJ62920-AEEA
170.01
CAD
CAAAJ62920-AE
CAAAJ62920-AK
Glycylglycine, 0.2 M Buffer Solution, pH 8.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 556-50-3
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00008130
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...