Chelerythrine chloride ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2-Dimethoxy-12-methyl-[1,3]dioxolo[4',5':4,5]benzo[1,2-c]phenanthridin-12-ium chloride
A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells
Purity: 99+%
CAS No.: 3895-92-9
Molecular Formula: C21H18ClNO4
Formula Weight: 383.82
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Solubility: Soluble in DMSO at 2mg/ml
Merck Reference: 14,2051
MDL No.: MFCD00060717
A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells
Formula:
C₂₁H₁₈ClNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00060717 CAS Number: 3895-92-9 EINECS: 223-444-9 Merck Index: 14,2051 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...