Sodium Borate, 0.5 M Buffer Solution, pH 8.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62902-AK J62902-AP
CAAAJ62902-AKEA 150.07 CAD
CAAAJ62902-AK CAAAJ62902-AP
Sodium Borate, 0.5 M Buffer Solution, pH 8.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 1330-43-4
Physical Form: Liquid
MDL No.: MFCD00081185

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical form
CAS No.
MDL No. MFCD00081185

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR