U-73122 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(6-(((8R,9S,13S,14S,17S)-3-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthren-17-yl)amino)hexyl)-1H-pyrrole-2,5-dione, 1-[6-[((17beta)-3-Methoxyestra-1,3,5[10]-trien-17-yl)amino]hexyl]pyrrole-2,5-dione

J62898-LB0 J62898-MA J62898-MCR
CAAAJ62898-LB0EA 432.71 CAD
CAAAJ62898-LB0 CAAAJ62898-MA CAAAJ62898-MCR
U-73122 95%
U-73122

Inhibitor of phospholipase C activation and inhibits the down regulation of muscarinic receptors


Crystalline solid


Soluble in DMSO, ethanol, or DMF at approx. 0.5mg/ml. May require heating or overnight mixing. Insoluble in water

Formula: C₂₉H₄₀N₂O₃
MW: 464.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00893825
CAS Number: 112648-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR