Aztreonam
Supplier: Thermo Scientific Chemicals
Synonyms:
2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulpho-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid, 2-((((Z)-1-(2-Aminothiazol-4-yl)-2-(((2S,3S)-2-methyl-4-oxo-1-sulphoazetidin-3-yl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid, 2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulfo-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid, 2-((((Z)-1-(2-Aminothiazol-4-yl)-2-(((2S,3S)-2-methyl-4-oxo-1-sulfoazetidin-3-yl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid
Synthetic monocyclic beta-lactam antibiotic. Inhibits cell wall synthesis in Gram-negative bacteria
Powder
Applications: Synthetic monocyclic beta-lactam antibiotic. Inhibits cell wall synthesis in Gram-negative bacteria
Formula:
C₁₃H₁₇N₅O₈S₂ MW: 435.43 g/mol Melting Pt: 227 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00072145 CAS Number: 78110-38-0 EINECS: 278-839-9 Merck Index: 14,925 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...