L-Methionine sulfoxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-Met(O)-OH, (2S)-2-Amino-4-(methylsulfinyl)butanoic acid, (2S)-2-Amino-4-(methylsulphinyl)butanoic acid
J62873-06
J62873-14
J62873-03
CAAAJ62873-06EA
403.99
CAD
CAAAJ62873-06
CAAAJ62873-14
CAAAJ62873-03
L-Methionine sulfoxide
L-Methionine sulphoxide
Powder
CAS No.: 3226-65-1
Molecular Formula: C5H11NO3S
Formula Weight: 165.21
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4129939
MDL No.: MFCD00063093
Formula:
C₅H₁₁NO₃S MW: 165.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00063093 CAS Number: 3226-65-1 EINECS: 221-758-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...