L(+)-Asparagine monohydrate cell culture reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O
J62869-22
J62869-36
J62869-A1
CAAAJ62869-22EA
212.35
CAD
CAAAJ62869-22
CAAAJ62869-36
CAAAJ62869-A1
L(+)-Asparagine monohydrate cell culture reagent
L(+)-Asparagine monohydrate
White to off-white
CAS No.: 5794-13-8
Molecular Formula: C4H8N2O3.H2O
Formula Weight: 150.14 (132.12anhy)
Physical Form: Powder
Appearance: White to off-white
Melting Point: 215-217°
Beilstein Reference: 1723527
MDL No.: MFCD00151038
Formula:
C₄H₈N₂O₃·1H₂O MW: 150.13 g/mol Melting Pt: 215…217 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151038 CAS Number: 5794-13-8 EINECS: 200-735-9 Merck Index: 14,00837 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...