Omeprazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole, 6-Methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulfinyl)-1H-benzo[d]imidazole

J62860-03 J62860-06
CAAAJ62860-03EA 134.71 CAD
CAAAJ62860-03 CAAAJ62860-06
Omeprazole 98%
Omeprazole

A proton pump inhibitor


Powder


Applications: A proton pump inhibitor

Formula: C₁₇H₁₉N₃O₃S
Melting Pt: 158…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00083192
CAS Number: 73590-58-6
Merck Index: 14,6845

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR