Phosphate-Buffered Saline (PBS, 10X), RNase free, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62851-AK J62851-AP
CAAAJ62851-AKEA 179.6 CAD
CAAAJ62851-AK CAAAJ62851-AP
Phosphate-Buffered Saline (PBS, 10X), RNase free, Thermo Scientific
Buffers Cell Culture Buffers

Liquid


Physical Form: Liquid
MDL No.: MFCD00131855

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR