Staurosporine ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J62837-LB0
J62837-MA
J62837-MCR
CAAAJ62837-LB0EA
915.23
CAD
CAAAJ62837-LB0
CAAAJ62837-MA
CAAAJ62837-MCR
Staurosporine ≥99%
Staurosporine
Broad spectrum protein kinase inhibitor
Purity: 99+%
CAS No.: 62996-74-1
Molecular Formula: C28H26N4O3
Formula Weight: 466.54
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: 240-243°
Solubility: Soluble in DMSO or ethanol
Beilstein Reference: 1060573
Merck Reference: 14,8802
MDL No.: MFCD00077402
Broad spectrum protein kinase inhibitor
Formula:
C₂₈H₂₆N₄O₃ Melting Pt: 240…243 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18252446 CAS Number: 62996-74-1 Merck Index: 14,8802 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...