Gentamicin sulfate 600 IU/mg

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gentamicin sulfate salt, NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

J62834-03 J62834-06 J62834-09
CAAAJ62834-03EA 135.91 CAD
CAAAJ62834-03 CAAAJ62834-06 CAAAJ62834-09
Gentamicin sulfate 600 IU/mg
Gentamicin sulphate

Gentamycin Sulfate is a broad-spectrum, aminoglycoside antibiotic used for cell culture which inhibits protein synthesis in sensitive organisms. Reported to have activity against gram-negative bacteria, Pseudomonas aeruginosa, gram-positive Staphylococcus aureus and some mycoplasmas. Reported to have low toxicity to mammalian cells and viruses at certain concentrations, useful for long-term tissue culture studies with viruses.


Composed of Gentamycin C1, Gentamycin C1a and Gentamycin C2.


Applications: Bacterial antibiotic that inhibits protein synthesis


Caution: Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS Number: 1405-41-0
EINECS: 215-778-9
Merck Index: 14,4392

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR