RNA sample loading buffer, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62832-S0
J62832-XF
CAAAJ62832-S0EA
242.85
CAD
CAAAJ62832-S0
CAAAJ62832-XF
RNA sample loading buffer, Thermo Scientific
Buffers
Electrophoresis Buffers
Loading Buffers
Liquid
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00213041
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...