Cimetidine ≥98%

Supplier: Thermo Scientific Chemicals
Danger

J62825-06 J62825-09 J62825-14
CAAAJ62825-06EA 248.94 CAD
CAAAJ62825-06 CAAAJ62825-09 CAAAJ62825-14
Cimetidine ≥98%
Cimetidine

H2 histamine receptor antagonist


Powder


Applications: H2 histamine receptor antagonist

Formula: C₁₀H₁₆N₆S
Melting Pt: 139…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00133296
CAS Number: 51481-61-9
EINECS: 257-232-2
Merck Index: 14,2279

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR