L(+)-Leucine cell culture reagent
Supplier: Thermo Scientific Chemicals
J62824-22
J62824-30
J62824-36
CAAAJ62824-22EA
260.37
CAD
CAAAJ62824-22
CAAAJ62824-30
CAAAJ62824-36
L(+)-Leucine cell culture reagent
L(+)-Leucine
Powder
CAS No.: 61-90-5
Molecular Formula: C6H13NO2
Formula Weight: 131.17
Physical Form: Powder
Density: 1.293
Appearance: White to off-white
Melting Point:
Formula:
C₆H₁₃NO₂ Melting Pt: >300 °C Density: 1.293 Storage Temperature: Ambient |
MDL Number:
MFCD00002617 CAS Number: 61-90-5 EINECS: 200-522-0 Merck Index: 14,5451 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...