Benzamidine hydrochloride hydrate, (H₂O 10-14%) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenylamidine hydrochloride hydrate, Benzene-1-carboximidamide hydrochloride hydrate, Benzenecarboximidamide hydrochloride hydrate, Benzamidinium chloride hydrate

J62823-06 J62823-09
CAAAJ62823-06EA 121.75 CAD
CAAAJ62823-06 CAAAJ62823-09
Benzamidine hydrochloride hydrate, (H₂O 10-14%) 99%
Benzamidine hydrochloride hydrate

Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases


Powder or beads


Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases

Formula: C₆H₅C(=NH)NH₂·HCl·H₂O
Melting Pt: 80…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00066285
CAS Number: 206752-36-5
EINECS: 216-795-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR