Candesartan 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylic acid, 1-2-1H-Tetrazol-5-yl-1-1-biphenyl-4-yl-methyl-2-ethoxy-1H-benzo-d-imidazole-7-carboxylic-acid, 1-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylic acid

J62818-03 J62818-MC
CAAAJ62818-03EA 778.24 CAD
CAAAJ62818-03 CAAAJ62818-MC
Candesartan 98%
Candesartan

Angiotensin II receptor antagonist


Powder


Applications: Angiotensin II receptor antagonist

Formula: C₂₄H₂₀N₆O₃
MW: 440.45 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00864463
CAS Number: 139481-59-7
Merck Index: 14,1739

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR