Nifedipine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J62811-06
J62811-14
J62811-22
CAAAJ62811-06EA
102.92
CAD
CAAAJ62811-06
CAAAJ62811-14
CAAAJ62811-22
Nifedipine 98%
Nifedipine
L-type calcium channel blocker; induces apoptosis in human glioblastoma cells
Powder
Applications: L-type calcium channel blocker; induces apoptosis in human glioblastoma cells
Formula:
C₁₇H₁₈N₂O₆ Melting Pt: 171…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00057326 CAS Number: 21829-25-4 EINECS: 244-598-3 Merck Index: 14,6528 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...