Sodium Bicarbonate, 1 M Buffer Solution, pH 9.4, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62808-AK J62808-AP
CAAAJ62808-AKEA 150.07 CAD
CAAAJ62808-AK CAAAJ62808-AP
Sodium Bicarbonate, 1 M Buffer Solution, pH 9.4, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 144-55-8
Physical Form: Liquid
MDL No.: MFCD00003528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR