Agarose Gel Loading Dye 6X for Electrophoresis, Ficoll Based, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62800-AB
J62800-AC
CAAAJ62800-ABEA
175.14
CAD
CAAAJ62800-AB
CAAAJ62800-AC
Agarose Gel Loading Dye 6X for Electrophoresis, Ficoll Based, Thermo Scientific
Stains
Electrophoresis Stains
Contains: 0.25% bromophenol blue, 0.25% xylene cyanol FF and 15% Ficoll in water