EDTA (ethylenediamine tetraacetic acid) 0.5 M in aqueous solution pH 8.0

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II

J62786-AK J62786-AP
CAAAJ62786-AKEA 121.41 CAD
CAAAJ62786-AK CAAAJ62786-AP
EDTA (ethylenediamine tetraacetic acid) 0.5 M in aqueous solution pH 8.0
EDTA (Ethylenediamine tetraacetic acid)

Liquid


CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Physical Form: Liquid Appearance: Colorless MDL No.: MFCD00003541

Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR