Orange G for electrophoresis

Supplier: Thermo Scientific Chemicals

Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10, Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulfonate

J62743-A1 J62743-14 J62743-22
CAAAJ62743-A1EA 389.88 CAD
CAAAJ62743-A1 CAAAJ62743-14 CAAAJ62743-22
Orange G for electrophoresis
Orange G

Collagen stain for connective tissue


Powder


Collagen stain for connective tissue

Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Melting Pt: 141 °C
Density: 0.45 to 0.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR