Paclitaxel ≥99.5%

Supplier: Thermo Scientific Chemicals
Danger

J62734-03 J62734-MC J62734-ME
CAAAJ62734-03EA 1691.69 CAD
CAAAJ62734-03 CAAAJ62734-MC CAAAJ62734-ME
Paclitaxel ≥99.5%
Paclitaxel

Promotes assembly and inhibits disassembly of microtubules (anti-cancer agent)


Crystalline solid


Applications: Promotes assembly and inhibits disassembly of microtubules (anti-cancer agent)

Formula: C₄₇H₅₁NO₁₄
Melting Pt: 213 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00869953
CAS Number: 33069-62-4
Merck Index: 14,6982

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR