Mefenamic Acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(2,3-Xylidino)benzoic acid, 2-((2,3-Dimethylphenyl)amino)benzoic acid, 2-(2,3-Dimethylphenylamino)benzoic acid, N-(2,3-Dimethylphenyl)anthranilic acid

J62705-22 J62705-14
CAAAJ62705-22EA 264.89 CAD
CAAAJ62705-22 CAAAJ62705-14
Mefenamic Acid 98%
Mefenamic acid

An non-steroidal anti-inflammatory drug with antiproliferative activity against human colon cancer cells


Powder


Applications: An non-steroidal anti-inflammatory drug with antiproliferative activity against human colon cancer cells

Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051721
CAS Number: 61-68-7
EINECS: 200-513-1
Merck Index: 14,5798

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR